Learn More
Med Vet International Ethicon Chromic Gut Suture, Size 4-0, SH-1, 27in, 36/Box
Catalog No. NC1985422
Encompass_Preferred
The Ethicon G181H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a SH taper point needle and has 36 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC1985422 Supplier Med Vet International Supplier No. G181H
The Ethicon G181H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a SH taper point needle and has 36 sutures in a box.