Learn More
Med Vet International Ethicon PDS II Suture, Size 5-0, P-3, 18in, 12/Box
Catalog No. NC1985425
Encompass_Preferred
The Ethicon Z463G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 18in. in length, has a size of 5-0, they are dyed Violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC1985425 Supplier Med Vet International Supplier No. Z463G
The Ethicon Z463G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 18in. in length, has a size of 5-0, they are dyed Violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.