Learn More
Med Vet International Ethicon Monocryl, 5-0 Poliglecaprone 25 Suture, 18in, P-3, Undyed, 12/Box
Catalog No. NC1656510
Encompass_Preferred
The Ethicon Y493G suture is a high-quality absorbable Poliglecaprone 25 undyed surgical suture. It measures 18in. in length, has a size of 5-0, they typically have an absorption rate of 90-120 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC1656510 Supplier Med Vet International Supplier No. Y493G
The Ethicon Y493G suture is a high-quality absorbable Poliglecaprone 25 undyed surgical suture. It measures 18in. in length, has a size of 5-0, they typically have an absorption rate of 90-120 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.