Learn More
Med Vet International Ethicon Vicryl Suture, Size 6-0, RB-1, 27in, 36/Box
Catalog No. NC1898431
Encompass_Preferred
The Ethicon J212H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 6-0, they are clear, and typically have an absorption rate of 56-70 days. This suture comes with a RB-1 taper point needle and has 36 sutures in a box.
Catalog No. NC1898431 Supplier Med Vet International Supplier No. J212HRL
The Ethicon J212H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 6-0, they are clear, and typically have an absorption rate of 56-70 days. This suture comes with a RB-1 taper point needle and has 36 sutures in a box.