Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 2-0, No Needle, 54in, Violet, 36/Box
Catalog No. 502830028
Encompass_Preferred
The Ethicon VCP615H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 54in in length, has a size of 2-0, and are violet. This suture comes with no needle and has 36 sutures in a box. Primary code substitute for discontinued code J615H
Catalog No. 50-283-0028 Supplier Med Vet International Supplier No. VCP615H
The Ethicon VCP615H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 54in in length, has a size of 2-0, and are violet. This suture comes with no needle and has 36 sutures in a box. Primary code substitute for discontinued code J615H