Learn More
Med Vet International Ethicon Coated Vicryl Suture, Size 8-0, TG140-8, 12in, 12/Box
Catalog No. NC1985432
Encompass_Preferred
The Ethicon J548G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 8-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a TG140-8 Spatula reverse cutting needle and has 12 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC1985432 Supplier Med Vet International Supplier No. J548G
The Ethicon J548G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 8-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a TG140-8 Spatula reverse cutting needle and has 12 sutures in a box.