Learn More
Med Vet International Ethicon Vicryl Suture, Size 6-0, PS-3, 18in, 12/Box
Catalog No. NC1656487
Encompass_Preferred
Vicryl J499G suture is a high-quality absorbable Polyglactin 910 surgical suture. Undyed and typically have an absorption rate of 56-70 days. This suture comes with a PS-3 Precision Point reverse cutting needle.
This item is not returnable. View return policy
Catalog No. NC1656487 Supplier Med Vet International Supplier No. J499G
Vicryl J499G suture is a high-quality absorbable Polyglactin 910 surgical suture. Undyed and typically have an absorption rate of 56-70 days. This suture comes with a PS-3 Precision Point reverse cutting needle.