Learn More
Med Vet International Ethicon Vicryl Suture, Size 7-0, TG140-8, 18in, 12/Box
Catalog No. NC1656525
Encompass_Preferred
The Ethicon J546G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 7-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a TG140-8 Spatula reverse cutting needle and has 12 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC1656525 Supplier Med Vet International Supplier No. J546G
The Ethicon J546G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 7-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a TG140-8 Spatula reverse cutting needle and has 12 sutures in a box.