Learn More
Med Vet International Oasis Chromic Gut Suture, Size 3-0, with NCT-1 Needle, 12/box, Veterinary Use Only
Supplier:  Med Vet International MV810V
Encompass_Preferred
The veterinary use Oasis Chromic Gut suture with needle is a natural absorbable suture that has a brown thread color and is extremely strong tensile strength, designed to provide secure knotting, and be more pliable and flexible. Size: 3-0. Needle Type: NCT-1, 36mm 1/2 Circle Taper Point. Length: 30in. Quantity: 12/box
Catalog No. 50-255-9407