Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 1, Double Armed LR LR, 30In, Black, 24/Box
Supplier:  Med Vet International 489T
Encompass_Preferred
The Ethicon 489T suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 30in in length, has a size of 1, and are dyed black to provide greater visibility while suturing. This suture comes with a Double Armed LR LR 75mm 3/8c Reverse Cutting Needle and has 24 sutures in a box.
Catalog No. 50-255-9716
 Restricted Item