Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 5-0, PS-3, 18in, 12/Box
Catalog No. 502092791
Encompass_Preferred
The Ethicon 1668G suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 5-0, and are dyed black to provide greater visibility while suturing. This suture comes with a PS-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-209-2791 Supplier Med Vet International Supplier No. 1668G
The Ethicon 1668G suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 5-0, and are dyed black to provide greater visibility while suturing. This suture comes with a PS-3 Precision Point reverse cutting needle and has 12 sutures in a box.