Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 5-0, PS-3, 18in, 36/Box
Catalog No. 502092792
Encompass_Preferred
The Ethicon 1668H suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 5-0, and are dyed black to provide greater visibility while suturing. This suture comes with a PS-3 Precision Point reverse cutting needle and has 36 sutures in a box.
Catalog No. 50-209-2792 Supplier Med Vet International Supplier No. 1668H
The Ethicon 1668H suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 5-0, and are dyed black to provide greater visibility while suturing. This suture comes with a PS-3 Precision Point reverse cutting needle and has 36 sutures in a box.