Learn More
TARGETMOL CHEMICALS INC SUTEZOLID 25MG
SDP
Catalog No. 502579191
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sutezolid (U-100480) is an oxazolidinone antimicrobial for the treatment of tuberculosis. purity: 99%
Catalog No. 50-257-9191 Supplier TARGETMOL CHEMICALS INC Supplier No. T741925MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sutezolid (U-100480) is an oxazolidinone antimicrobial for the treatment of tuberculosis. purity: 99%