Learn More
Med Vet International Ethicon Monocryl Poliglecaprone 25 Suture, Size 2-0, CT, 36In, Violet, 36/Box
Supplier:  Med Vet International Y357H
Encompass_Preferred
The Ethicon Y357H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 36in in length, has a size of 2-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a CT 40mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9288