Learn More
Med Vet International Ethicon Monocryl Poliglecaprone 25 Suture, Size 3-0, RB-1, 27In, Violet, 36/Box
Supplier:  Med Vet International Y305H
Encompass_Preferred
The Ethicon Y305H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 27in in length, has a size of 3-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9293