Learn More
Med Vet International Ethicon Monocryl Poliglecaprone 25 Suture, Size 3-0, SH, 27In, Undyed, 36/Box
Supplier:  Med Vet International Y416H
Encompass_Preferred
The Ethicon Y416H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 27in in length, has a size of 3-0, and are undyed. This suture comes with a SH 26mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9285