Learn More
Med Vet International Ethicon PDS Plus Antibacterial Polydioxanone Suture, Size 2-0, SH, 27In, Violet, 36/Box
Supplier:  Med Vet International PDP317H
Encompass_Preferred
The Ethicon PDP317H suture is a high-quality absorbable Pds Plus Antibacterial Polydioxanone suture. It measures 27in in length, has a size of 2-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a SH 26mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9328
 Restricted Item