Learn More
Med Vet International Ethicon PDS Plus Antibacterial Polydioxanone Suture, Size 3-0, SH-1, 27In, Violet, 36/Box
Supplier:  Med Vet International PDP311H
Encompass_Preferred
The Ethicon PDP311H suture is a high-quality absorbable Pds Plus Antibacterial Polydioxanone suture. It measures 27in in length, has a size of 3-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a SH-1 22mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9331