Learn More
Med Vet International Ethicon PDS II Suture, Size 6-0, CP-1, 18in, 12/Box
Catalog No. 502092805
Encompass_Preferred
The Ethicon Z487G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 18in. in length, has a size of 6-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a CP-1 Prime reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-209-2805 Supplier Med Vet International Supplier No. Z487G
The Ethicon Z487G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 18in. in length, has a size of 6-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a CP-1 Prime reverse cutting needle and has 12 sutures in a box.