Learn More
Med Vet International Ethicon Permahand Silk Suture, Size 0, CT-1, 30In, Black, 36/Box
Supplier:  Med Vet International 424H
Encompass_Preferred
The Ethicon 424H suture is a high-quality non-absorbable Permahand Silk suture. It measures 30in in length, has a size of 0, and are dyed black to provide greater visibility while suturing. This suture comes with a CT-1 36mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9717