Learn More
Med Vet International Ethicon Permahand Silk Suture, Size 2-0, RB-1, 30In, Black, 36/Box
Supplier:  Med Vet International K873H
Encompass_Preferred
The Ethicon K873H suture is a high-quality non-absorbable Permahand Silk suture. It measures 30in in length, has a size of 2-0, and are dyed black to provide greater visibility while suturing. This suture comes with a RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9566