Learn More
Med Vet International Oasis Silk Suture, Size 4-0, With Nfs-1 Needle, 12/Box
Catalog No. 502559409
Encompass_Preferred
The Oasis Silk suture with needle is a braided non-absorbable suture. This sterile surgical suture is composed of an organic protein called fibroin. This protein is derived from the domesticated species Bombyx mori (B. mori) of the family Bombycidae. This line of sutures has FDA approved black dye for enhanced visibility.
This item is not returnable. View return policy
Catalog No. 50-255-9409 Supplier Med Vet International Supplier No. MV629
The Oasis Silk suture with needle is a braided non-absorbable suture. This sterile surgical suture is composed of an organic protein called fibroin. This protein is derived from the domesticated species Bombyx mori (B. mori) of the family Bombycidae. This line of sutures has FDA approved black dye for enhanced visibility.