Learn More
Med Vet International Oasis Silk Suture, Size 4-0, with NFS-2 Needle, 12/box
Supplier:  Med Vet International MV683
Encompass_Preferred
The Oasis Silk suture with needle is a braided non-absorbable suture. This sterile surgical suture is composed of an organic protein called fibroin. This protein is derived from the domesticated species Bombyx mori (B. mori) of the family Bombycidae. This line of sutures has FDA approved black dye for enhanced visiblity.Size: 4-0. Needle Type: NFS-2, 19mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. 50-255-9408