Learn More
Med Vet International Ethicon Permahand Silk Suture, Size 6-0, G-1, 18in, 12/Box
Catalog No. 502092811
Encompass_Preferred
The Ethicon 786G suture is a high-quality non-absorbable Silk surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a G-1 reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-209-2811 Supplier Med Vet International Supplier No. 786G
The Ethicon 786G suture is a high-quality non-absorbable Silk surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a G-1 reverse cutting needle and has 12 sutures in a box.