Learn More
Med Vet International Ethicon Permahand Silk Suture, Size 6-0, P-1, 18IN, 12/Box
Catalog No. 502092812
Encompass_Preferred
The Ethicon 639G suture is a high-quality non-absorbable Silk surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a P-1 Precision Point reverse cutting needle and has 12 sutures in a box.
This item is not returnable. View return policy
Catalog No. 50-209-2812 Supplier Med Vet International Supplier No. 639G
 Restricted Item
Add to Cart
Add to Cart
The Ethicon 639G suture is a high-quality non-absorbable Silk surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a P-1 Precision Point reverse cutting needle and has 12 sutures in a box.