Learn More
Med Vet International Oasis Chromic Gut Cassette, Size 3-0, Length of 50M, Each
Catalog No. 502522420
Encompass_Preferred
The veterinary use Oasis Chromic Gut suture cassette is a natural absorbable suture that has a brown thread color and has extremely strong tensile strength, designed to provide secure knotting, and be more pliable and flexible. Size: 3-0. Length: 50M.
Catalog No. 50-252-2420 Supplier Med Vet International Supplier No. CG30
The veterinary use Oasis Chromic Gut suture cassette is a natural absorbable suture that has a brown thread color and has extremely strong tensile strength, designed to provide secure knotting, and be more pliable and flexible. Size: 3-0. Length: 50M.