Learn More
Med Vet International Suture Cassette Rack, Holds 5 Cassettes, J0091R5, Each
Catalog No. 5000585276
Encompass_Preferred
Stainless steel. Heavy-duty model. Cassettes lock into place and cannot slip out. Holds 5 cassettes
Catalog No. 50-005-85276 Supplier Med Vet International Supplier No. JOR91R5
Stainless steel. Heavy-duty model. Cassettes lock into place and cannot slip out. Holds 5 cassettes