Learn More
Med Vet International Ethicon Chromic Gut Suture, Size 4-0, FS-2, 27in, 36/Box
Catalog No. 501180547
Encompass_Preferred
The Ethicon 635H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a FS-2 reverse cutting needle and has 36 sutures in a box.
Catalog No. 50-118-0547 Supplier Med Vet International Supplier No. 635H
The Ethicon 635H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a FS-2 reverse cutting needle and has 36 sutures in a box.