Learn More
Med Vet International Ethicon Chromic Gut Suture, Size 0, CT-2, 27in, 36/Box
Catalog No. 502522489
Encompass_Preferred
The Ethicon 884H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a CT-2 taper point needle and has 36 sutures in a box.
Catalog No. 50-252-2489 Supplier Med Vet International Supplier No. 884H
The Ethicon 884H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a CT-2 taper point needle and has 36 sutures in a box.