Learn More
Med Vet International Chromic Gut Suture, 0, Ncp-1, Veterinary Use, 12/Box
Catalog No. 502829438
Encompass_Preferred
Natural absorbable gut suture, Excellent knot security, Very high tensile strength, Improved pliability and flexibility, Composition: Natural Purified Collagen, Absorption Type: Absorbable, Structure: Natural, Thread Color: Brown
Catalog No. 50-282-9438 Supplier Med Vet International Supplier No. MV817V
Natural absorbable gut suture, Excellent knot security, Very high tensile strength, Improved pliability and flexibility, Composition: Natural Purified Collagen, Absorption Type: Absorbable, Structure: Natural, Thread Color: Brown