Learn More
Med Vet International Ethicon Chromic Gut Suture, Size 2-0, CT-1, 27in, 36/Box
Catalog No. 502522513
Encompass_Preferred
The Ethicon 811H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 2-0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-252-2513 Supplier Med Vet International Supplier No. 811H
The Ethicon 811H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 2-0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.