Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 2, LR, 20in, Black, 24/Box
Catalog No. 502522535
Encompass_Preferred
The Ethicon 460T suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 20in in length, has a size of 2, and are dyed black to provide greater visibility while suturing. This suture comes with a LR 75mm 3/8c Reverse Cutting Needle and has 24 sutures in a box.
Catalog No. 50-252-2535 Supplier Med Vet International Supplier No. 460T
The Ethicon 460T suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 20in in length, has a size of 2, and are dyed black to provide greater visibility while suturing. This suture comes with a LR 75mm 3/8c Reverse Cutting Needle and has 24 sutures in a box.