Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 3-0, PC-5, 18in, Black, 12/Box
Catalog No. 502522577
Encompass_Preferred
The Ethicon 1893G suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 18in in length, has a size of 3-0, and are dyed black to provide greater visibility while suturing. This suture comes with a PC-5 19mm 3/8c Conventional Cutting Needle and has 12 sutures in a box.
Catalog No. 50-252-2577 Supplier Med Vet International Supplier No. 1893G
The Ethicon 1893G suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 18in in length, has a size of 3-0, and are dyed black to provide greater visibility while suturing. This suture comes with a PC-5 19mm 3/8c Conventional Cutting Needle and has 12 sutures in a box.