Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 4-0, FS-2, 18in, 12/Box
Catalog No. 501180576
Encompass_Preferred
The Ethicon 662G suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 4-0, and are dyed black to provide greater visibility while suturing. This suture comes with a FS-2 reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-118-0576 Supplier Med Vet International Supplier No. 662G
The Ethicon 662G suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 4-0, and are dyed black to provide greater visibility while suturing. This suture comes with a FS-2 reverse cutting needle and has 12 sutures in a box.