Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 6-0, P-1, 18in, Black, 12/Box
Catalog No. 502522523
Encompass_Preferred
The Ethicon 689G suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 18in in length, has a size of 6-0, and are undyed. This suture comes with a P-1 11mm 3/8c Reverse Cutting Needle and has 12 sutures in a box.
Catalog No. 50-252-2523 Supplier Med Vet International Supplier No. 689G
The Ethicon 689G suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 18in in length, has a size of 6-0, and are undyed. This suture comes with a P-1 11mm 3/8c Reverse Cutting Needle and has 12 sutures in a box.