Learn More
Med Vet International Ethicon Ethilon Nylon Suture 6-0, P-3 needle, 18in, Black Qty/Box, 12
Catalog No. 502522579
Encompass_Preferred
The Ethicon 1698G suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2579 Supplier Med Vet International Supplier No. 1698G
The Ethicon 1698G suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.