Learn More
Fine Science Tools Suture Tying Forceps
SDP
Catalog No. NC9903040
Encompass
Product: Suture Tying Forceps - 10cm/ Tip Dimensions: 0.5 x 0.5mm/ Platform Length: 5mm/ Alloy: Stainless Steel/ Length: 10cm/ Unit: Each. A pin inside the shanks limits the applied pressure.
Catalog No. NC9903040 Supplier Fine Science Tools Supplier No. 1802510
Product: Suture Tying Forceps - 10cm/ Tip Dimensions: 0.5 x 0.5mm/ Platform Length: 5mm/ Alloy: Stainless Steel/ Length: 10cm/ Unit: Each. A pin inside the shanks limits the applied pressure.