Learn More
Med Vet International Oasis Monofyl Suture, Size 3-0, with NFS-2 Needle, 12/Box, Veterinary Use Only
Catalog No. 502522248
Encompass_Preferred
The Oasis veterinary use Monofyl suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ Monocryl®. The Monofyl suture has an absorption rate between 91 and 119 days. Size: 3-0. Needle Type: NFS-2, 19mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. 50-252-2248 Supplier Med Vet International Supplier No. MVY923V
 Restricted Item
Add to Cart
Add to Cart
The Oasis veterinary use Monofyl suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ Monocryl®. The Monofyl suture has an absorption rate between 91 and 119 days. Size: 3-0. Needle Type: NFS-2, 19mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box