Learn More
Med Vet International Monofilament Synthetic Absorbable Suture, 4-0 Thread Size, Cutting Needle, Each
Catalog No. 502522258
Encompass_Preferred
Monofilament Synthetic Absorbable Suture. Absorbable sutures break down in the body and are often used internally. Monofilament suture material is made of a single strand. This structure is relatively more resistant to harboring microorganisms. Reverse cutting needles have their cutting surfaces on the convex surface. Reverse cutting needles have a reduced risk of cutting through tissue than conventional cutting needles. These types of needles are typically used in ophthalmic and cosmetic surgery.
Catalog No. 50-252-2258 Supplier Med Vet International Supplier No. MSA40C
Monofilament Synthetic Absorbable Suture. Absorbable sutures break down in the body and are often used internally. Monofilament suture material is made of a single strand. This structure is relatively more resistant to harboring microorganisms. Reverse cutting needles have their cutting surfaces on the convex surface. Reverse cutting needles have a reduced risk of cutting through tissue than conventional cutting needles. These types of needles are typically used in ophthalmic and cosmetic surgery.