Learn More
Med Vet International Ethicon Monocryl Suture, Size 0, CT-1, 27in, 36/Box
Catalog No. 502522130
Encompass_Preferred
The Ethicon Y340H suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 27in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 90-120 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-252-2130 Supplier Med Vet International Supplier No. Y340H
The Ethicon Y340H suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 27in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 90-120 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.