Learn More
Med Vet International Ethicon Monocryl Suture, Size 3-0, PS-2, 18in, 12/Box
Catalog No. 502522122
Encompass_Preferred
The Ethicon Y497G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 18in. in length, has a size of 3-0, they typically have an absorption rate of 90-120 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2122 Supplier Med Vet International Supplier No. Y497G
The Ethicon Y497G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 18in. in length, has a size of 3-0, they typically have an absorption rate of 90-120 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.