Learn More
Med Vet International Ethicon Monocryl Suture, Size 3-0, SH, 27in, 36/Box
Catalog No. 502522133
Encompass_Preferred
The Ethicon Y316H suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 90-120 days. This suture comes with a SH taper point needle and has 36 sutures in a box.
Catalog No. 50-252-2133 Supplier Med Vet International Supplier No. Y316H
The Ethicon Y316H suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 90-120 days. This suture comes with a SH taper point needle and has 36 sutures in a box.