Learn More
Med Vet International Ethicon Monocryl Suture 4-0, P-3 Needle, 18in, 12/Box
Catalog No. 502522124
Encompass_Preferred
The Ethicon Y494G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 18in. in length, has a size of 4-0, they typically have an absorption rate of 90-120 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2124 Supplier Med Vet International Supplier No. Y494G
The Ethicon Y494G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 18in. in length, has a size of 4-0, they typically have an absorption rate of 90-120 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.