Learn More
Med Vet International Ethicon Monocryl 4-0 Size: 4-0 Needle: PS-2 Length: 18in Color: Undyed Qty/bx: 12
Catalog No. 502522123
Encompass_Preferred
The Ethicon Y496G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 18in. in length, has a size of 4-0, is undyed and they typically have an absorption rate of 90-120 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2123 Supplier Med Vet International Supplier No. Y496G
The Ethicon Y496G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 18in. in length, has a size of 4-0, is undyed and they typically have an absorption rate of 90-120 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.