Learn More
Med Vet International Ethicon Monocryl Suture, Size 5-0, P-1, 18in, undyed 12/Box
Catalog No. 502522125
Encompass_Preferred
The Ethicon Y490G suture is a high-quality absorbable Poliglecaprone 25 undyed surgical suture. It measures 18in. in length, has a size of 5-0, they typically have an absorption rate of 90-120 days. This suture comes with a P-1 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2125 Supplier Med Vet International Supplier No. Y490G
The Ethicon Y490G suture is a high-quality absorbable Poliglecaprone 25 undyed surgical suture. It measures 18in. in length, has a size of 5-0, they typically have an absorption rate of 90-120 days. This suture comes with a P-1 Precision Point reverse cutting needle and has 12 sutures in a box.