Learn More
Med Vet International Ethicon Monocryl, 5-0 Poliglecaprone 25 Suture, 18in, P-3, 12/Box
Catalog No. 502522127
Encompass_Preferred
The Ethicon Y463G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 18in. in length, has a size of 5-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 90-120 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2127 Supplier Med Vet International Supplier No. Y463G
The Ethicon Y463G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures 18in. in length, has a size of 5-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 90-120 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.