Learn More
Med Vet International Ethicon Monocryl Plus Antibacterial Suture, Size 3-0, Ps-2, 27in, Undyed, 36/Box
Catalog No. 502831496
Encompass_Preferred
The Ethicon MCP427H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 27in in length, has a size of 3-0, and are undyed. This suture comes with a PS-2 19mm 3/8c Cutting Edge Prime Reverse Needle and has 36 sutures in a box. Primary code substitute for discontinued code H822H
Catalog No. 50-283-1496 Supplier Med Vet International Supplier No. MCP427H
The Ethicon MCP427H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 27in in length, has a size of 3-0, and are undyed. This suture comes with a PS-2 19mm 3/8c Cutting Edge Prime Reverse Needle and has 36 sutures in a box. Primary code substitute for discontinued code H822H