Learn More
Med Vet International Ethicon Monocryl Plus Antibacterial Suture, Size 4-0, P-3, 18in, Undyed, 12/Box
Catalog No. 502831158
Encompass_Preferred
The Ethicon MCP494G suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 18in in length, has a size of 4-0, and are undyed. This suture comes with a P-3 13mm 3/8c Cutting Edge Prime Reverse Needle and has 12 sutures in a box. Primary code substitute for discontinued code 1654G
Catalog No. 50-283-1158 Supplier Med Vet International Supplier No. MCP494G
The Ethicon MCP494G suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 18in in length, has a size of 4-0, and are undyed. This suture comes with a P-3 13mm 3/8c Cutting Edge Prime Reverse Needle and has 12 sutures in a box. Primary code substitute for discontinued code 1654G