Learn More
Med Vet International Ethicon Monocryl Plus Antibacterial Suture, Size 5-0, Ps-2, 18in, Undyed, 12/Box
Catalog No. 502830991
Encompass_Preferred
The Ethicon MCP495G suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 18in in length, has a size of 5-0, and are undyed. This suture comes with a PS-2 19mm 3/8c Cutting Edge Prime Reverse Needle and has 12 sutures in a box. Primary code substitute for discontinued code 634G
Catalog No. 50-283-0991 Supplier Med Vet International Supplier No. MCP495G
The Ethicon MCP495G suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 18in in length, has a size of 5-0, and are undyed. This suture comes with a PS-2 19mm 3/8c Cutting Edge Prime Reverse Needle and has 12 sutures in a box. Primary code substitute for discontinued code 634G