Learn More
Med Vet International Ethicon Monocryl Poliglecaprone 25 Suture, Size 4-0, RB-1, 27in, Undyed, 36/Box
Catalog No. 502522136
Encompass_Preferred
The Ethicon Y214H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 27in in length, has a size of 4-0, and are undyed. This suture comes with a RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2136 Supplier Med Vet International Supplier No. Y214H
The Ethicon Y214H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 27in in length, has a size of 4-0, and are undyed. This suture comes with a RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box.